2,4-Difluorobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

D2839-25G D2839-5G
TCD2839-25GEA 120.3 USD
TCD2839-25G TCD2839-5G
2,4-Difluorobenzaldehyde ≥98.0% (by GC)
2,4-Difluorobenzaldehyde

CAS Number: 1550-35-2
MDL Number: MFCD00010326
Molecular Formula: C7H4F2O
Molecular Weight: 142.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 55

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Flash Pt: 55 °C
MDL Number: MFCD00010326
CAS Number: 1550-35-2
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69cd863bdfbe5aaedf2120aa8e664fd46f53576f&vid=49885215fb985566ed2ab3c3e6ba38ba26894014&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR