2,2-Dimethylcaproic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2-Dimethylhexanoic acid

D2809-1G D2809-5G
TCD2809-1GEA 119 USD
TCD2809-1G TCD2809-5G
2,2-Dimethylcaproic acid ≥98.0% (by GC, titration analysis)
2,2-Dimethylcaproic acid

CAS Number: 813-72-9
MDL Number: MFCD00965097
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 0.91

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 122 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00965097
CAS Number: 813-72-9
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4029928ffa25a72554016ba8b2bc99e42143c7e4&vid=4029928ffa25a72554016ba8b2bc99e42143c7e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR