Diphenyl(methyl)sulfonium tetrafluoroborate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 10504-60-6
MDL Number: MFCD00051861
Molecular Formula: C13H13BF4S
Molecular Weight: 288.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 66
Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis), Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Methyldiphenylsulfonium Tetrafluoroborate
Formula:
C₁₃H₁₃BF₄S Melting Pt: 66 °C |
MDL Number:
MFCD00051861 CAS Number: 10504-60-6 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...