3,5-Difluoroaniline ≥98.0% (by GC)
Supplier: TCI America
D2665-25G
D2665-5G
TCD2665-25GEA
197
USD
TCD2665-25G
TCD2665-5G
3,5-Difluoroaniline ≥98.0% (by GC)
3,5-Difluoroaniline
CAS Number: 372-39-4
MDL Number: MFCD00007763
Molecular Formula: C6H5F2N
Molecular Weight: 129.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 82
Melting point (°C): 39
Flash Point (°C): 75
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 82 °C Melting Pt: 39 °C Flash Pt: 75 °C |
MDL Number:
MFCD00007763 CAS Number: 372-39-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...