2,4-Dimethylthiophenol ≥96.0% (by GC)

Supplier: TCI America

D2648-25G D2648-5G
TCD2648-25GEA 153.35 USD
TCD2648-25G TCD2648-5G
2,4-Dimethylthiophenol ≥96.0% (by GC)
2,4-Dimethylthiophenol

CAS Number: 13616-82-5
MDL Number: MFCD00010019
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 58
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2,4-Dimethylthiophenol, 4-Mercapto-m-xylene, m-Xylene-4-thiol

Formula: C₈H₁₀S
Boiling Pt: 214 °C
Density: 1.03 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00010019
CAS Number: 13616-82-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8fcd4f30d31defaed597306c94b3901a4e4795e&vid=a1c8c19502eff66c24795536796ae5e8537787d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR