3,5-Difluorobenzyl bromide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α-Bromo-3,5-difluorotoluene

D2631-25G D2631-5G
TCD2631-25GEA 250 USD
TCD2631-25G TCD2631-5G
3,5-Difluorobenzyl bromide ≥98.0% (by GC)
3,5-Difluorobenzyl bromide

CAS Number: 141776-91-2
MDL Number: MFCD00010304
Molecular Formula: C7H5BrF2
Molecular Weight: 207.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Flash Point (°C): 81
Specific Gravity (20/20): 1.62


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-3,5-difluorotoluene

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 65 °C
Density: 1.62 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00010304
CAS Number: 141776-91-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=913dfd07500f4ea131557cf72d2527f0b07a91ab&vid=913dfd07500f4ea131557cf72d2527f0b07a91ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR