α-Bromo-3,4-difluorotoluene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3,4-Difluorobenzyl bromide, 4-(Bromomethyl)-1,2-difluorobenzene
D2627-25G
D2627-5G
TCD2627-25GEA
322.2
USD
TCD2627-25G
TCD2627-5G
α-Bromo-3,4-difluorotoluene ≥98.0% (by GC)
α-Bromo-3,4-difluorotoluene
CAS Number: 85118-01-0
MDL Number: MFCD00009889
Molecular Formula: C7H5BrF2
Molecular Weight: 207.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 79
Specific Gravity (20/20): 1.63
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-3,4-difluorotoluene
Formula:
C₇H₅BrF₂ MW: 207.02 g/mol Boiling Pt: 61 °C Density: 1.63 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00009889 CAS Number: 85118-01-0 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...