2,3-Dichloroquinoxaline ≥98.0% (by GC)

Supplier: TCI America

D2622-25G D2622-5G
TCD2622-25GEA 146 USD
TCD2622-25G TCD2622-5G
2,3-Dichloroquinoxaline ≥98.0% (by GC)
2,3-Dichloroquinoxaline

CAS Number: 2213-63-0
MDL Number: MFCD00006720
Molecular Formula: C8H4Cl2N2
Molecular Weight: 199.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 153

Formula: C₈H₄Cl₂N₂
Melting Pt: 153 °C
MDL Number: MFCD00006720
CAS Number: 2213-63-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edf5e32dd30f11a7da6dd5f02c73ec7467e3031b&vid=edf5e32dd30f11a7da6dd5f02c73ec7467e3031b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR