3,4'-Dichloropropiophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2613-25G D2613-5G
TCD2613-25GEA 244 USD
TCD2613-25G TCD2613-5G
3,4'-Dichloropropiophenone ≥98.0% (by GC, titration analysis)
3,4'-Dichloropropiophenone

CAS Number: 3946-29-0
MDL Number: MFCD00000992
Molecular Formula: C9H8Cl2O
Molecular Weight: 203.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 81

Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
Melting Pt: 51 °C
Flash Pt: 81 °C
MDL Number: MFCD00000992
CAS Number: 3946-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=325dcd2a4edb592f9222050955681d35eba26157&vid=325dcd2a4edb592f9222050955681d35eba26157&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR