3,5-Dimethoxybenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2609-25G D2609-5G
TCD2609-25GEA 226 USD
TCD2609-25G TCD2609-5G
3,5-Dimethoxybenzoyl chloride ≥98.0% (by GC, titration analysis)
3,5-Dimethoxybenzoyl chloride

CAS Number: 17213-57-9
MDL Number: MFCD00000676
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 29

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00000676
CAS Number: 17213-57-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eed740e1732c8ef57783e4a7db7269cc2ed61a3&vid=8eed740e1732c8ef57783e4a7db7269cc2ed61a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR