4,6-Dichloro-2-(methylthio)pyrimidine ≥98.0% (by GC)

Supplier: TCI America

D2533-25G D2533-5G
TCD2533-25GEA 91 USD
TCD2533-25G TCD2533-5G
4,6-Dichloro-2-(methylthio)pyrimidine ≥98.0% (by GC)
4,6-Dichloro-2-(methylthio)pyrimidine

CAS Number: 6299-25-8 MDL Number: MFCD00006086 Molecular Formula: C5H4Cl2N2S Molecular Weight: 195.06 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 136 Melting point (°C): 42 Flash Point (°C): 110 Storage Temperature: 0-10°C

Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
Boiling Pt: 136 °C
Melting Pt: 42 °C
Flash Pt: 110 °C
MDL Number: MFCD00006086
CAS Number: 6299-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a044a1f502ac0c85cad0e778dbdca5637f49a780&vid=a044a1f502ac0c85cad0e778dbdca5637f49a780&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR