Ethyl difluoroacetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Difluoroacetic ethyl ester

D2498-25G D2498-5G
TCD2498-25GEA 152.7 USD
TCD2498-25G TCD2498-5G
Ethyl difluoroacetate ≥98.0% (by GC)
Ethyl difluoroacetate

CAS Number: 454-31-9
MDL Number: MFCD00013578
Molecular Formula: C4H6F2O2
Molecular Weight: 124.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 24
Specific Gravity (20/20): 1.18


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Difluoroacetic Acid Ethyl Ester

Formula: C₄H₆F₂O₂
MW: 124.09 g/mol
Boiling Pt: 98 °C
Density: 1.18 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00013578
CAS Number: 454-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b40d58e4ca74fcdcd5ce4ab52d150d23c9f36670&vid=b40d58e4ca74fcdcd5ce4ab52d150d23c9f36670&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR