4,6-Dichloropyrimidine ≥98.0% (by GC)

Supplier: TCI America

D2488-25G D2488-5G
TCD2488-25GEA 48 USD
TCD2488-25G TCD2488-5G
4,6-Dichloropyrimidine ≥98.0% (by GC)
4,6-Dichloropyrimidine

CAS Number: 1193-21-1
MDL Number: MFCD00006109
Molecular Formula: C4H2Cl2N2
Molecular Weight: 148.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00006109
CAS Number: 1193-21-1
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcf987dd17b9f07505491625cc09259a462996f6&vid=dcf987dd17b9f07505491625cc09259a462996f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR