3,4-Difluorophenol ≥98.0% (by GC)

Supplier: TCI America

D2476-25G D2476-5G
TCD2476-25GEA 191 USD
TCD2476-25G TCD2476-5G
3,4-Difluorophenol ≥98.0% (by GC)
3,4-Difluorophenol

CAS Number: 2713-33-9
MDL Number: MFCD00010315
Molecular Formula: C6H4F2O
Molecular Weight: 130.09
Purity/Analysis Method: >98.0% (GC)
Flash Point (°C): 57
Freezing point (°C): 35

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Flash Pt: 57 °C
MDL Number: MFCD00010315
CAS Number: 2713-33-9
UN: 2921
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d4ddbd0e9ef47c05aaf0d821fe016e039e0f01f&vid=718c1d2b0d7bbc43bdd26f0599d35c58dffd9337&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR