2,3-Difluorophenol ≥98.0% (by GC)

Supplier: TCI America

D2453-25G D2453-5G
TCD2453-25GEA 158 USD
TCD2453-25G TCD2453-5G
2,3-Difluorophenol ≥98.0% (by GC)
2,3-Difluorophenol

CAS Number: 6418-38-8
MDL Number: MFCD00010262
Molecular Formula: C6H4F2O
Molecular Weight: 130.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 54
Melting point (°C): 40
Flash Point (°C): 56

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 54 °C
Melting Pt: 40 °C
Flash Pt: 56 °C
MDL Number: MFCD00010262
CAS Number: 6418-38-8
UN: 2921
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d21e517ef3a067040b08a12c501d3ad16a7dbee&vid=3860ccd952fae7f4b7accbc06e85f328b1c8b7c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR