3,4-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2451-25G D2451-5G
TCD2451-25GEA 294 USD
TCD2451-25G TCD2451-5G
3,4-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)
3,4-Dichlorobenzylamine

CAS Number: 102-49-8
MDL Number: MFCD00008114
Molecular Formula: C7H7Cl2N
Molecular Weight: 176.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 1.33

Formula: C₇H₇Cl₂N
Boiling Pt: 140 °C
Density: 1.33 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008114
CAS Number: 102-49-8
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb84bd56e3a30189fa394d082be90041724be49c&vid=cb84bd56e3a30189fa394d082be90041724be49c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR