2,6-Dimethyl-3,5-heptanedione ≥97.0% (by GC)

Supplier: TCI America

D2413-25G D2413-5G
TCD2413-25GEA 420 USD
TCD2413-25G TCD2413-5G
2,6-Dimethyl-3,5-heptanedione ≥97.0% (by GC)
2,6-Dimethyl-3,5-heptanedione

CAS Number: 18362-64-6
MDL Number: MFCD00015040
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Specific Gravity (20/20): 0.91


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Diisobutyrylmethane

Formula: C₉H₁₆O₂
MW: 156.22 g/mol
Boiling Pt: 80 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00015040
CAS Number: 18362-64-6
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2aa80411e9493c8a2344dd596ec1198ad6610a8&vid=d15ec76a5e9e2d287bc62019da141c6983443fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR