Dicyclohexylmethane-4,4'-diisocyanate (mixture of isomers) ≥90.0% (by GC)

Supplier: TCI America

D2380-25G D2380-500G
TCD2380-25GEA 26.46 USD
TCD2380-25G TCD2380-500G
Dicyclohexylmethane-4,4'-diisocyanate (mixture of isomers) ≥90.0% (by GC)
Dicyclohexylmethane-4,4'-diisocyanate (mixture of isomers)

CAS Number: 5124-30-1
MDL Number: MFCD00015405
Molecular Formula: C15H22N2O2
Molecular Weight: 262.35
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 113
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4,4'-Methylenebis(cyclohexyl Isocyanate)

Formula: C₁₅H₂₂N₂O₂
Boiling Pt: 168 °C
Density: 1.07 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00015405
CAS Number: 5124-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1c8d0942dea5e816df5251181ba445e7ef35af4&vid=a1c8d0942dea5e816df5251181ba445e7ef35af4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR