cis-2,6-Dimethylpiperazine ≥98.0%
Supplier: TCI America
D2360-25G
D2360-500G
TCD2360-25GEA
64.9
USD
TCD2360-25G
TCD2360-500G
cis-2,6-Dimethylpiperazine ≥98.0%
cis-2,6-Dimethylpiperazine
CAS Number: 21655-48-1
MDL Number: MFCD00005956
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 113
Flash Point (°C): 45
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol Boiling Pt: 160 °C Melting Pt: 113 °C Flash Pt: 45 °C |
MDL Number:
MFCD07772435 CAS Number: 21655-48-1 UN: 1325 ADR: 4.1 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...