Diphenyliodonium nitrate
Supplier: TCI America
D2357-25G
D2357-5G
TCD2357-25GEA
235
USD
TCD2357-25G
TCD2357-5G
Diphenyliodonium nitrate
Diphenyliodonium nitrate
CAS Number: 722-56-5
MDL Number: MFCD00031704
Molecular Formula: C12H10INO3
Molecular Weight: 343.12
Form: Crystal
Color: White
Formula:
C₁₂H₁₀INO₃ MW: 343.12 |
MDL Number:
MFCD00031704 CAS Number: 722-56-5 UN: 3087 ADR: 5.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...