3,4-Difluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

D2341-25G D2341-5G
TCD2341-25GEA 146.5 USD
TCD2341-25G TCD2341-5G
3,4-Difluorobenzonitrile ≥98.0% (by GC)
3,4-Difluorobenzonitrile

CAS Number: 64248-62-0
MDL Number: MFCD00011666
Molecular Formula: C7H3F2N
Molecular Weight: 139.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 50
Flash Point (°C): 69


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-1,2-difluorobenzene

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 95 °C
Melting Pt: 50 °C
Flash Pt: 69 °C
MDL Number: MFCD00011666
CAS Number: 64248-62-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7e9e78508b1cc4b1e2abf67cbc341117711c7e0&vid=cac9c5c582695c6316f521fe8729ff5a8cc0ab09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR