2,6-Difluorotoluene ≥99.0% (by GC)

Supplier: TCI America

D2338-1G D2338-5G
TCD2338-1GEA 19 USD
TCD2338-1G TCD2338-5G
2,6-Difluorotoluene ≥99.0% (by GC)
2,6-Difluorotoluene

CAS Number: 443-84-5
MDL Number: MFCD00043898
Molecular Formula: C7H6F2
Molecular Weight: 128.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 10
Specific Gravity (20/20): 1.13

Formula: C₇H₆F₂
Boiling Pt: 112 °C
Density: 1.13 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00043898
CAS Number: 443-84-5
UN: 2388
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84c5120ec939fbd8d1c81fdda214b218bf6dd808&vid=c26f7cea8366a0bdf8b14406544fb3730dff36bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR