α-Chloro-2,6-difluorotoulene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(Chloromethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl chloride, 1-(Chloromethyl)-2,6-difluorobenzene

D2337-25G D2337-5G
TCD2337-25GEA 111 USD
TCD2337-25G TCD2337-5G
α-Chloro-2,6-difluorotoulene ≥98.0% (by GC)
α-Chloro-2,6-difluorotoulene

CAS Number: 697-73-4
MDL Number: MFCD00191346
Molecular Formula: C7H5ClF2
Molecular Weight: 162.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 38


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Chloro-2,6-difluorotoluene

Formula: C₇H₅ClF₂
MW: 162.57 g/mol
MDL Number: MFCD00191346
CAS Number: 697-73-4
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67278f893714346e17430c62b41ba9e53094b8e7&vid=67278f893714346e17430c62b41ba9e53094b8e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR