1,3-Dichloro-1,1,3,3-tetramethyldisiloxane ≥97.0% (by GC)
Supplier: TCI America
D2334-25G
D2334-5G
TCD2334-25GEA
173.25
USD
TCD2334-25G
TCD2334-5G
1,3-Dichloro-1,1,3,3-tetramethyldisiloxane ≥97.0% (by GC)
1,3-Dichloro-1,1,3,3-tetramethyldisiloxane
CAS Number: 2401-73-2
MDL Number: MFCD00018088
Molecular Formula: C4H12Cl2OSi2
Molecular Weight: 203.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 43
Specific Gravity (20/20): 1.04
Formula:
C₄H₁₂Cl₂OSi₂ MW: 203.21 g/mol Boiling Pt: 138 °C Density: 1.04 (20 °C) Flash Pt: 43 °C |
MDL Number:
MFCD00018088 CAS Number: 2401-73-2 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...