Diphenylsilanediol ≥98.0% (by GC)

Supplier: TCI America

D2316-250G D2316-25G
TCD2316-250GEA 169 USD
TCD2316-250G TCD2316-25G
Diphenylsilanediol ≥98.0% (by GC)
Diphenylsilanediol

CAS Number: 947-42-2
MDL Number: MFCD00002101
Molecular Formula: C12H12O2Si
Molecular Weight: 216.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147
Flash Point (°C): 53


Other Product Names / Synonyms: Dihydroxydiphenylsilane

Formula: C₁₂H₁₂O₂Si
Melting Pt: 147 °C
Flash Pt: 53 °C
MDL Number: MFCD00002101
CAS Number: 947-42-2
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c3b9899b8dc95fa5731fb45efa15904960cf956&vid=4c3b9899b8dc95fa5731fb45efa15904960cf956&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR