4,4'-Diiodobiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Diiodo-1,1'-biphenyl

D2315-250G D2315-25G
TCD2315-250GEA 233 USD
TCD2315-250G TCD2315-25G
4,4'-Diiodobiphenyl ≥98.0%
4,4'-Diiodobiphenyl

CAS Number: 3001-15-8
MDL Number: MFCD00001057
Molecular Formula: C12H8I2
Molecular Weight: 406.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 208

Formula: C₁₂H₈I₂
MW: 406 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00001057
CAS Number: 3001-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42a9410f18f4580b303e4d8f0e2cdc1f9eff38e7&vid=42a9410f18f4580b303e4d8f0e2cdc1f9eff38e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR