5,6-Dihydro-2H-pyran-2-one ≥95.0% (by GC)

Supplier: TCI America

D2261-1G D2261-25G D2261-5G
TCD2261-1GEA 83 USD
TCD2261-1G TCD2261-25G TCD2261-5G
5,6-Dihydro-2H-pyran-2-one ≥95.0% (by GC)
5,6-Dihydro-2H-pyran-2-one

CAS Number: 3393-45-1
MDL Number: MFCD00010439
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 46
Specific Gravity (20/20): 1.15

Formula: C₅H₆O₂
Boiling Pt: 117 °C
Density: 1.15 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00010439
CAS Number: 3393-45-1
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12d7580115e5c706d1a17f4126060265125e57cc&vid=12d7580115e5c706d1a17f4126060265125e57cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR