Dimethylphenylsilane ≥97.0%
Supplier: TCI America
Synonyms:
Dimethyl(phenyl)silane
D2196-25ML
D2196-5ML
TCD2196-25MLEA
131.55
USD
TCD2196-25ML
TCD2196-5ML
Dimethylphenylsilane ≥97.0%
Dimethylphenylsilane
CAS Number: 766-77-8
MDL Number: MFCD00008257
Molecular Formula: C8H12Si
Molecular Weight: 136.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 35
Specific Gravity (20/20): 0.88
Formula:
C₈H₁₂Si MW: 136.27 g/mol Boiling Pt: 158 °C Density: 0.88 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00008257 CAS Number: 766-77-8 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...