4,4'-Difluorobiphenyl

Supplier: TCI America

Synonyms: 4,4-Difluoro-1,1-biphenyl, PFB No. 15

D2181-1G D2181-5G
TCD2181-1GEA 33.5 USD
TCD2181-1G TCD2181-5G
4,4'-Difluorobiphenyl
4,4'-Difluorobiphenyl

CAS Number: 398-23-2
MDL Number: MFCD00000349
Molecular Formula: C12H8F2
Molecular Weight: 190.19
Form: Crystal
Color: White
Boiling point (°C): 255
Melting point (°C): 89

Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 255 °C
Melting Pt: 89 °C
MDL Number: MFCD00000349
CAS Number: 398-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c62aaee297c178ad9a48e6c34cea34540bf80027&vid=c62aaee297c178ad9a48e6c34cea34540bf80027&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR