4,5-Dimethylthiazole ≥98.0% (by GC)

Supplier: TCI America

D2142-25G D2142-5G
TCD2142-25GEA 85 USD
TCD2142-25G TCD2142-5G
4,5-Dimethylthiazole ≥98.0% (by GC)
4,5-Dimethylthiazole

CAS Number: 3581-91-7 MDL Number: MFCD00005336 Molecular Formula: C5H7NS Molecular Weight: 113.18 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 159 Flash Point (°C): 51 Specific Gravity (20/20): 1.08

Formula: C₅H₇NS
Boiling Pt: 159 °C
Density: 1.08 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00005336
CAS Number: 3581-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12c4ec9183944c20d610dd5fe01b1407a7c1cb36&vid=12c4ec9183944c20d610dd5fe01b1407a7c1cb36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR