Dimethoxy(methyl)silane ≥97.0% (by GC)

Supplier: TCI America

D2100-25ML D2100-100ML
TCD2100-25MLEA 193.76 USD
TCD2100-25ML TCD2100-100ML
Dimethoxy(methyl)silane ≥97.0% (by GC)
Dimethoxy(methyl)silane

CAS Number: 16881-77-9
MDL Number: MFCD00048019
Molecular Formula: C3H10O2Si
Molecular Weight: 106.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): -9
Specific Gravity (20/20): 0.86

Formula: C₃H₁₀O₂Si
Boiling Pt: 61 °C
Density: 0.86 (20 °C)
Flash Pt: –9 °C
MDL Number: MFCD00048019
CAS Number: 16881-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79aaa4aa107c4a3eb59ce698c83a491e89434e40&vid=79aaa4aa107c4a3eb59ce698c83a491e89434e40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR