3-(Dicyanomethylidene)indan-1-one ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
2-(3-Oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile
D2054-1G
D2054-5G
TCD2054-1GEA
145.4
USD
TCD2054-1G
TCD2054-5G
3-(Dicyanomethylidene)indan-1-one ≥98.0% (by total nitrogen basis)
3-(Dicyanomethylidene)indan-1-one
CAS Number: 1080-74-6
MDL Number: MFCD00143117
Molecular Formula: C12H6N2O
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 228
Formula:
C₁₂H₆N₂O MW: 194.19 g/mol Melting Pt: 228 °C |
MDL Number:
MFCD00143117 CAS Number: 1080-74-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...