2,6-Difluorophenol ≥98.0% (by GC)

Supplier: TCI America

D2015-25G D2015-5G
TCD2015-25GEA 264.7 USD
TCD2015-25G TCD2015-5G
2,6-Difluorophenol ≥98.0% (by GC)
2,6-Difluorophenol

CAS Number: 28177-48-2
MDL Number: MFCD00002158
Molecular Formula: C6H4F2O
Molecular Weight: 130.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 60
Melting point (°C): 45
Flash Point (°C): 58

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 60 °C
Melting Pt: 45 °C
Flash Pt: 58 °C
MDL Number: MFCD00002158
CAS Number: 28177-48-2
UN: 2921
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84dea3ae9c48c3ac335c0a42d81a7ee08a78c387&vid=157db0c0d861ff66f2884750a7ef37aceae198f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR