2,4-Difluorobenzylamine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(2,4-Difluorophenyl)methanamine
D1996-1G
D1996-5G
TCD1996-1GEA
21
USD
TCD1996-1G
TCD1996-5G
2,4-Difluorobenzylamine ≥98.0% (by GC)
2,4-Difluorobenzylamine
CAS Number: 72235-52-0
MDL Number: MFCD00010142
Molecular Formula: C7H7F2N
Molecular Weight: 143.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 57
Flash Point (°C): 68
Specific Gravity (20/20): 1.22
Formula:
C₇H₇F₂N MW: 143.14 g/mol Boiling Pt: 57 °C Density: 1.22 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00010142 CAS Number: 72235-52-0 UN: 2735 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...