1-Bromo-2,6-difluorobenzene ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 64248-56-2
MDL Number: MFCD00009894
Molecular Formula: C6H3BrF2
Molecular Weight: 192.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 53
Freezing point (°C): 20
Specific Gravity (20/20): 1.72
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,6-Difluorobromobenzene
Formula:
C₆H₃BrF₂ Boiling Pt: 137 °C Density: 1.72 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00009894 CAS Number: 64248-56-2 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...