1,2-Dibromo-4,5-difluorobenzene ≥97.0% (by GC)

Supplier: TCI America

D1913-25G D1913-5G
TCD1913-25GEA 105 USD
TCD1913-25G TCD1913-5G
1,2-Dibromo-4,5-difluorobenzene ≥97.0% (by GC)
1,2-Dibromo-4,5-difluorobenzene

CAS Number: 64695-78-9
MDL Number: MFCD00009890
Molecular Formula: C6H2Br2F2
Molecular Weight: 271.89
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 44
Freezing point (°C): 32

Formula: C₆H₂Br₂F₂
Flash Pt: 44 °C
MDL Number: MFCD00009890
CAS Number: 64695-78-9
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d71346556bbd79106730b1622e97bb85153ee347&vid=1c6a5d8f5bb615494d898d815782c2fe27a6e0ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR