1-Bromo-2,4-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Difluorobromobenzene

D1909-25G D1909-500G
TCD1909-25GEA 28 USD
TCD1909-25G TCD1909-500G
1-Bromo-2,4-difluorobenzene ≥98.0% (by GC)
1-Bromo-2,4-difluorobenzene

CAS Number: 348-57-2
MDL Number: MFCD00000330
Molecular Formula: C6H3BrF2
Molecular Weight: 192.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 51
Specific Gravity (20/20): 1.71


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2,4-Difluorobromobenzene

Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 146 °C
Density: 1.71 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00000330
CAS Number: 348-57-2
UN: 2924
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5754b3483b84eff93aaec847daa6a3ee626b1cd&vid=e81553b3f20ba54cad61c27483c2ff4f0bea4631&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR