(Diethylamino)sulphur trifluoride ≥90.0%
Supplier: TCI America
Synonyms:
DAST
D1868-100G
D1868-25G
D1868-5G
TCD1868-100GEA
1158
USD
TCD1868-100G
TCD1868-25G
TCD1868-5G
(Diethylamino)sulphur trifluoride ≥90.0%
(Diethylamino)sulphur trifluoride
[Fluorinating Reagent]
CAS Number: 38078-09-0
MDL Number: MFCD00000363
Molecular Formula: C4H10F3NS
Molecular Weight: 161.19
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid
Boiling point (°C): 32
Flash Point (°C): 23
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Glycosylation, Fluorinating Agents, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: DAST, Sulfur Trifluoride Diethylamine Complex
Formula:
C₄H₁₀F₃NS MW: 161.19 g/mol Boiling Pt: 32 °C Density: 1.23 (20 °C) Flash Pt: 23 °C |
MDL Number:
MFCD00000363 CAS Number: 38078-09-0 UN: 2920 ADR: 8 I |
|
Specification Test Results
Purity/Analysis Method | >90.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...