2,4-Difluoroanisole ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Difluoro-1-methoxybenzene

D1849-10G D1849-25G
TCD1849-10GEA 62 USD
TCD1849-10G TCD1849-25G
2,4-Difluoroanisole ≥99.0% (by GC)
2,4-Difluoroanisole

CAS Number: 452-10-8
MDL Number: MFCD00010731
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 53
Flash Point (°C): 51
Specific Gravity (20/20): 1.24

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 53 °C
Density: 1.24 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00010731
CAS Number: 452-10-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73112d9e07e5fd15fb49a73b1cdb9e4b64bb53ce&vid=65d26373585cfefaa944daa1a7e1eac6605c2539&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR