2,6-Difluorobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1847-25G D1847-5G
TCD1847-25GEA 234.3 USD
TCD1847-25G TCD1847-5G
2,6-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
2,6-Difluorobenzylamine

CAS Number: 69385-30-4
MDL Number: MFCD00010144
Molecular Formula: C7H7F2N
Molecular Weight: 143.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 65
Specific Gravity (20/20): 1.22

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 177 °C
Density: 1.22 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00010144
CAS Number: 69385-30-4
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c083cdad8709ba4e9bc083e71e74263036f9dca&vid=0c083cdad8709ba4e9bc083e71e74263036f9dca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR