Dimethylketene methyl trimethylsilyl acetal ≥95.0% (by GC)

Supplier: TCI America

D1789-25ML D1789-5ML
TCD1789-25MLEA 163.83 USD
TCD1789-25ML TCD1789-5ML
Dimethylketene methyl trimethylsilyl acetal ≥95.0% (by GC)
Dimethylketene methyl trimethylsilyl acetal

CAS Number: 31469-15-5
MDL Number: MFCD00010232
Molecular Formula: C8H18O2Si
Molecular Weight: 174.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 52
Flash Point (°C): 30
Specific Gravity (20/20): 0.86

Formula: C₈H₁₈O₂Si
Boiling Pt: 52 °C
Density: 0.86 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00010232
CAS Number: 31469-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e33d8c326097fddc1f2c973c12598ef126739a6&vid=9e33d8c326097fddc1f2c973c12598ef126739a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR