3,5-Dimethylisoxazole ≥99.0% (by GC)

Supplier: TCI America

D1767-250ML D1767-25ML
TCD1767-250MLEA 165.3 USD
TCD1767-250ML TCD1767-25ML
3,5-Dimethylisoxazole ≥99.0% (by GC)
3,5-Dimethylisoxazole

CAS Number: 300-87-8 MDL Number: MFCD00003156 Molecular Formula: C5H7NO Molecular Weight: 97.12 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 141 Flash Point (°C): 31 Specific Gravity (20/20): 0.99

Formula: C₅H₇NO
MW: 97.12 g/mol
Boiling Pt: 141 °C
Density: 0.99 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00003156
CAS Number: 300-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48a28f1b2ced829873e56965ed77d23b82a7b45&vid=e48a28f1b2ced829873e56965ed77d23b82a7b45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR