Desyl chloride ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Chloro-2-phenylacetophenone
D1745-25G
D1745-5G
TCD1745-25GEA
90
USD
TCD1745-25G
TCD1745-5G
Desyl chloride ≥98.0% (by GC)
Desyl chloride
CAS Number: 447-31-4
MDL Number: MFCD00000858
Molecular Formula: C14H11ClO
Molecular Weight: 230.69
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68
Other Product Names / Synonyms: alpha-Chlorodeoxybenzoin, Desyl Chloride
Formula:
C₁₄H₁₁ClO MW: 230.69 g/mol Melting Pt: 68 °C |
MDL Number:
MFCD00000858 CAS Number: 447-31-4 UN: 3261 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...