2,6-Difluoropyridine ≥98.0% (by GC)

Supplier: TCI America

D1741-25G D1741-5G
TCD1741-25GEA 113 USD
TCD1741-25G TCD1741-5G
2,6-Difluoropyridine ≥98.0% (by GC)
2,6-Difluoropyridine

CAS Number: 1513-65-1
MDL Number: MFCD00006227
Molecular Formula: C5H3F2N
Molecular Weight: 115.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 33
Specific Gravity (20/20): 1.27

Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 125 °C
Density: 1.27 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00006227
CAS Number: 1513-65-1
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ddf93ae2a0bd8c3e5f50e53d8e44c83f1aef056&vid=8ddf93ae2a0bd8c3e5f50e53d8e44c83f1aef056&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR