2,4-Dichlorobenzylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1740-25G D1740-5G
TCD1740-25GEA 430 USD
TCD1740-25G TCD1740-5G
2,4-Dichlorobenzylamine ≥98.0% (by titrimetric analysis)
2,4-Dichlorobenzylamine

CAS Number: 95-00-1
MDL Number: MFCD00008109
Molecular Formula: C7H7Cl2N
Molecular Weight: 176.04
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.33

Formula: C₇H₇Cl₂N
Density: 1.33 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008109
CAS Number: 95-00-1
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81af2780fc32a2835190eeba101283188b9abcbb&vid=81af2780fc32a2835190eeba101283188b9abcbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR