Dicyclohexano-18-crown-6 (mixture of isomers) ≥98.0% (by GC)

Supplier: TCI America

D1668-1G D1668-5G
TCD1668-1GEA 45 USD
TCD1668-1G TCD1668-5G
Dicyclohexano-18-crown-6 (mixture of isomers) ≥98.0% (by GC)
Dicyclohexano-18-crown-6 (mixture of isomers)

CAS Number: 16069-36-6
MDL Number: MFCD00005099
Molecular Formula: C20H36O6
Molecular Weight: 372.50
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 344
Melting point (°C): 51
Flash Point (°C): 113

Formula: C₂₀H₃₆O₆
Boiling Pt: 344 °C
Melting Pt: 51 °C
Flash Pt: 113 °C
MDL Number: MFCD00005099
CAS Number: 16069-36-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR