DFDNB (1,5-Difluoro-2,4-dinitrobenzene) ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
DFDNB, 1,5-Difluoro-2,4-dinitrobenzene
D1649-25G
D1649-5G
TCD1649-25GEA
169
USD
TCD1649-25G
TCD1649-5G
DFDNB (1,5-Difluoro-2,4-dinitrobenzene) ≥97.0% (by GC)
DFDNB (1,5-Difluoro-2,4-dinitrobenzene)
CAS Number: 327-92-4
MDL Number: MFCD00007052
Molecular Formula: C6H2F2N2O4
Molecular Weight: 204.09
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 74
Formula:
C₆H₂F₂N₂O₄ MW: 204.09 g/mol Melting Pt: 74 °C |
MDL Number:
MFCD00007052 CAS Number: 327-92-4 UN: 2811 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...