1,4-Difluorobenzene ≥99.0% (by GC)
Supplier: TCI America
D1628-25G
D1628-5G
TCD1628-25GEA
30
USD
TCD1628-25G
TCD1628-5G
1,4-Difluorobenzene ≥99.0% (by GC)
1,4-Difluorobenzene
CAS Number: 540-36-3
MDL Number: MFCD00000344
Molecular Formula: C6H4F2
Molecular Weight: 114.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Melting point (°C): -13
Flash Point (°C): -5
Specific Gravity (20/20): 1.17
Formula:
C₆H₄F₂ MW: 114.09 g/mol Boiling Pt: 88 °C Melting Pt: -13 °C Density: 1.17 (20 °C) Flash Pt: -5 °C |
MDL Number:
MFCD00000344 CAS Number: 540-36-3 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...