1,2-Difluorobenzene ≥98.0% (by GC)
Supplier: TCI America
D1627-25G
D1627-5G
TCD1627-25GEA
82
USD
TCD1627-25G
TCD1627-5G
1,2-Difluorobenzene ≥98.0% (by GC)
1,2-Difluorobenzene
CAS Number: 367-11-3
MDL Number: MFCD00000284
Molecular Formula: C6H4F2
Molecular Weight: 114.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 2
Specific Gravity (20/20): 1.17
Formula:
C₆H₄F₂ MW: 114.09 g/mol Boiling Pt: 91 °C Density: 1.17 (20 °C) Flash Pt: 2 °C |
MDL Number:
MFCD00000284 CAS Number: 367-11-3 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...