2,5-Dimethylthiophene ≥98.0% (by GC)

Supplier: TCI America

D1591-25ML D1591-5ML
TCD1591-25MLEA 83 USD
TCD1591-25ML TCD1591-5ML
2,5-Dimethylthiophene ≥98.0% (by GC)
2,5-Dimethylthiophene

CAS Number: 638-02-8
MDL Number: MFCD00005452
Molecular Formula: C6H8S
Molecular Weight: 112.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 24
Specific Gravity (20/20): 0.99

Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 136 °C
Density: 0.99 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00005452
CAS Number: 638-02-8
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e43b81bacb81482d1c22b211a5a09da10b85aa01&vid=e43b81bacb81482d1c22b211a5a09da10b85aa01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR